Learn More
ALKBH6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567311
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GMGMLNLEIGG DAGGRIGCKE LVLMEEQDAR VPALEPFRVE QAPPVIYYVP DFISKEEEEY LLRQVFNAPK PKWTQLS Highest antigen sequence identity to the following orthologs - mouse 68%, rat 67%.
Probable dioxygenase that requires molecular oxygen, alpha-ketoglutarate and iron.Specifications
ALKBH6 | |
Polyclonal | |
Unconjugated | |
ALKBH6 | |
ABH6; AI839550; alkB homolog 6; alkB, alkylation repair homolog 6; alkB, alkylation repair homolog 6 (E. coli); ALKBH6; Alkylated DNA repair protein alkB homolog 6; alkylation repair homolog 6; alpha-ketoglutarate-dependent dioxygenase alkB homolog 6; C130099A02; probable alpha-ketoglutarate-dependent dioxygenase ABH6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84964 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3KRA9 | |
ALKBH6 | |
Recombinant Human ALKBH6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only