Learn More
AGK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562795
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IVKGETVPLD VLQIKGEKEQ PVFAMTGLRW GSFRDAGVKV SKYWYLGPLK IKAAHFFSTL KEWPQTHQAS ISYTGPTERP PNEPEETPVQ RPSLYRRILR RLASYWAQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.Specifications
AGK | |
Polyclonal | |
Unconjugated | |
AGK | |
2610037M15Rik; 6720408I04Rik; acylglycerol kinase; acylglycerol kinase, mitochondrial; Agk; AI465370; CATC5; CTRCT38; FLJ10842; hAGK; hsMuLK; MTDPS10; Mulk; multiple substrate lipid kinase; multi-substrate lipid kinase; RGD1562046 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55750 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53H12 | |
AGK | |
Recombinant protein corresponding to Human AGK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only