missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, high gelling temperature, Molecular Biology Reagent

£78.09 - £181.91

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15400637
SDS View Documents
Thermo Scientific Chemicals
J66704.14
25 g
£78.09
25g
In Stock
to see stock.
Add to basket
 
15410637
SDS View Documents
Thermo Scientific Chemicals
J66704.22
100 g
£181.91
100g
In Stock
to see stock.
Add to basket
 
Description

Description

High gel strength allows gel formation at low concentrations. Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High gel strength allows gel formation at low concentrations
  • Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications
Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
White
MFCD00081294
9012-36-6
Hygroscopic; Moisture sensitive
Agarose, high gelling temperature, Molecular Biology Reagent
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-731-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.