Learn More
Thermo Scientific Chemicals Agarose D1-LE, molecular biology grade
£58.89 - £715.56
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387019
|
Thermo Scientific Chemicals
H26855.14 |
25 g |
£58.89
25g |
In Stock
Log in to see stock. |
|||||
11397019
|
Thermo Scientific Chemicals
H26855.22 |
100 g |
£176.88
100g |
In Stock
Log in to see stock. |
|||||
11307029
|
Thermo Scientific Chemicals
H26855.36 |
500 g |
£715.56
500g |
In Stock
Log in to see stock. |
|||||
Description
Agarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAgarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.
Solubility
Soluble in water.
Notes
Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
9012-36-6 | |
MFCD00081294 |
Specifications
9012-36-6 | |
MFCD00081294 | |
Molecular Biology |
Hygroscopic | |
Soluble in water. | |
Agarose D1-LE |
Safety and Handling
missing translation for 'einecsNumber' : 232-731-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.