missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adropin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560031
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SADVDSLSES SPNSSPGPCP EKAPPPQKPS HEGSYLLQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Adropin is involved in the regulation of glucose homestasis and lipid metabolism. It is a secreted protein encoded by the Energy Homeostasis Associated gene (ENHO) found in the brain and liver. In the liver, the protein has been found to increase in response to high-fat foods and decrease after fasting. The protein seems to play a role in controlling the activity of other metabolic genes, specifically those involved in the production of lipids from carbohydrates.Specifications
Adropin | |
Polyclonal | |
Unconjugated | |
ENHO | |
1110065P19Rik; 2310040A07Rik; Adropin; adropin-like; C9orf165; energy homeostasis associated; energy homeostasis-associated protein; ENHO; GAAI470; LOC100912292; RGD1565232; UNQ470; UNQ470/PRO830 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375704 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWT2 | |
ENHO | |
Recombinant protein corresponding to Human Adropin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only