Learn More
3M™ Adflo™ Heavy Duty Lithium Ion Battery
3M™ Adflo™ Heavy Duty Lithium Ion Battery makes the Adflo™ Powered Air Respirator even more lightweight and balanced for greater comfort during the full shift.
Brand: 3M™ 7000144539
Code : NEW
Additional Details : Weight : 4.32000kg Transport : UN number : 3480 Chem class : 9
Description
- Fully charges in 5.5 hours. Expected operating time is 10-13 hours with new particle filter, new fully-charged battery and a standard airflow
- Batteries may be stored on the charger, fully charged, ready for immediate use
- For use with the 3M™ Adflo™ Powered Air Respirator
Specifications
Turbo: approx. 10 to 13 hr. |