Learn More
3M™ Adflo™ Battery Charger (Li-ion)
3M™ Adflo™ Battery Charger (Li-ion) is a replacement charger suitable for use with Adflo™ Powered Air Respirator.
Brand: 3M™ 7000144537
Code : NEW
Additional Details : Weight : 11.00000kg
Description
- Replacement battery charger for Adflo Respirator Li-ion battery
- Will charge Adflo standard battery in 4.5 hours or Adflo heavy-duty battery in 5.5 hours
- For use with the 3M™ Adflo™ Powered Air Respirator