missing translation for 'onlineSavingsMsg'
Learn More

Manutan Add bay for longspan shelving, chipboard, 2000mm x 1800mm x 1000mm

Brand:  Manutan 245M035A

Encompass_Preferred

Product Code. 12847689

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Create a run of shelving as long as you require. Quick, easy boltless assembly - frames and beams simply clip together. Beams in grey and upright frames in blue with the level heights adjusting every 62. 5mm.

  • Three shelves
  • Shelf capacity UDL: 504kg
  • Dimensions (H x W x D): 2000mm x 1800mm x 1000mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.