Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
Brand:  Eppendorf™ 5910708009
245.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15873222
  • £250.00 / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microtube
11 mm
S-4x400 Bucket
26
26 x 1.5/2.0 mL Microtubes
39 mm
26
2/Pk.