Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
Brand:  Eppendorf™ 5910711000
245.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15803232
  • £250.00 / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round Bottom
12 mm
S-4x400 Bucket
17
17 x 12 x 75 mm Round Bottom Tubes
144 mm
17
2/Pk.