missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13911-1-AP-20UL
This item is not returnable.
View return policy
Description
Centaurin alpha-1 (ADAP1, ArfGAP With Dual PH Domains 1) is a Protein Coding gene. Among its related pathways are B Cell Receptor Signaling Pathway (sino) and Arf6 signaling events. Gene Ontology (GO) annotations related to this gene include GTPase activator activity and inositol 1,3,4,5 tetrakisphosphate binding. An important paralog of this gene is ADAP2.Specifications
ADAP1 | |
Polyclonal | |
Unconjugated | |
ADAP1 | |
ADAP1, ArfGAP with dual PH domains 1, CENTA1, Centaurin alpha 1, Cnt a1, GCS1L, p42IP4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11033, 171097, 231821 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75689 | |
Adap1 | |
ADAP1 Fusion Protein Ag4898 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |