missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550540
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total ADAMTS6 protein.
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The encoded preproprotein is proteolytically processed to generate the mature enzyme. Expression of this gene may be regulated by the cytokine TNF-alpha. [provided by RefSeq, Mar 2016].Specifications
ADAMTS6 | |
Polyclonal | |
Unconjugated | |
ADAMTS6 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 6; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 6; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 6; ADAM metallopeptidase with thrombospondin type 1 motif, 6; ADAMs; ADAM-TS 6; ADAMTS6; ADAM-TS6; ADAMTS-6; metalloendopeptidases | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11174 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UKP5 | |
ADAMTS6 | |
Synthetic peptide corresponding to a region derived from internal residues of human ADAM metallopeptidase with thrombospondin type 1 motif, 6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |