Learn More
ADAM33 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564312
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NSAGDAHGNC GQDSEGHFLP CAGRDALCGK LQCQGGKPSL LAPHMVPVDS TVHLDGQEVT CRGALALPSA QLD Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein is a type I transmembrane protein implicated in asthma and bronchial hyperresponsiveness. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifications
ADAM33 | |
Polyclonal | |
Unconjugated | |
Adam33 | |
a disintegrin and metallopeptidase domain 33; a disintegrin and metalloprotease 33; a disintegrin and metalloprotease domain 33; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 33; ADAM; ADAM 33; ADAM metallopeptidase domain 33; ADAM33; Adaml; ADAMs; C20orf153; disintegrin and metalloproteinase domain-containing protein 33; disintegrin and reprolysin metalloproteinase family protein; DJ964F7.1; DKFZp434K0521; FLJ35308; FLJ36751; HGNC:15478; metalloendopeptidases; MGC149823; MGC71889; RP23-228H14.4; RP5-964F7.2; UNQ873/PRO1891 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80332 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZ11 | |
Adam33 | |
Recombinant protein corresponding to Human ADAM33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only