missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Activated Carbon Filter

Aids in ventilation in safety storage cabinets.

Brand:  Asecos™ 32952

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15664844

  • £496.08 / Each
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Activated carbon
  • Available in different sizes
  • Accessory item
Specifications

Specifications

270 x 270 x 230 mm
9 kg
Filter
CX, SLX models
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers