missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ ActivArmr™ 97-011 Series 3/4 Dipped Medium Duty Gloves

Product Code. 15529210
Click to view available options
Size:
8
9
10
11

Protects workers from the elements. Ansell ActivArmr™ 97-011 Series 97-011 Series 3/4 Dipped Medium Duty Gloves keeps your hands warm in cold conditions while keeping the entire system breathable.

  • Industrial medium duty glove
  • 2 ply approach keeps yours hands warm in cold conditions while keeping the entire system breathable
  • 3/4 nitrile dip provides protection from direct wind, liquid and cold impact
  • Sandy nitrile finish ensures secure grip in multiple applications
  • Soft inner layer of acrylic terry and the nitrile coating provides comfort and dexterity
  • Ergonomically designed shape ensures perfect fit and decreases hand fatigue
  • Contrasting black coating against the bright orange liner offers extra visibility in low or poor light conditions

Applications:

  • Loading trucks and vehicles
  • Product shipping, transport and delivery
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003, A1:2009 and EN 388:2003
Coating Material Nitrile
Color Black, Orange
Exterior Finish 3/4 Dipped
Latex Free No
Material Nitrile
Product Type Medium-duty Industrial Gloves
Size 8
Glove Cuff Style Knit
Liner Material Acrylic Polyester
Packaging Type 12 inners in carton, 6 pairs in inner carton
Resistant To Cuts
Series 97-011
Length (Metric) 240 to 270 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'