missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (40%, 39:1), Thermo Scientific Chemicals

£73.87 - £117.59

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 39:1
Physical Form Liquid
Concentration 40% soln
Color Colorless
UN Number UN3426
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495579
View Documents
Thermo Scientific Alfa Aesar
J63323.AP
500 mL
£73.87
500mL
Please to purchase this item. Need a web account? Register with us today!
15405589
View Documents
Thermo Scientific Alfa Aesar
J63323.K2
1 L
£117.59
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

Solubility
Fully miscible in water.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

Acrylamide/Bisacrylamide 39:1
40% soln
UN3426
Liquid
Colorless
Fully miscible in water.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c

H302+H332-H315-H317-H319-H340-H350-H361f-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.