missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with Nylon Membrane
Versatile filter for both aqueous and solvent-based sample filtration.
Brand: Cytiva (formerly Pall Lab) AP-4436
119.70 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- Excellent chemical compatibility with esters, bases, and alcohols
- Saves time because no prewetting is required
Specifications
Nylon | |
Non-sterile | |
Polypropylene | |
25 mm |
0.2 μm | |
3.9 cm2 | |
50/Pk. |