Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with Nylon Membrane
Versatile filter for both aqueous and solvent-based sample filtration.
Brand: Cytiva (formerly Pall Lab) AP-4786
Additional Details : Weight : 1.13400kg
Description
- Excellent chemical compatibility with esters, bases, and alcohols
- Saves time because no prewetting is required
Specifications
Glass Fiber, Nylon | |
Non-sterile | |
Polypropylene | |
25/AutoPack Tubes |
0.2 μm | |
3.9 cm2 | |
200/Cs. |