Learn More
Acetyl chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals
£23.80 - £75.78
Chemical Identifiers
CAS | 75-36-5 |
---|---|
Molecular Formula | C2H3ClO |
Molecular Weight (g/mol) | 78.50 |
MDL Number | MFCD00000719 |
InChI Key | WETWJCDKMRHUPV-UHFFFAOYSA-N |
Synonym | ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 |
PubChem CID | 6367 |
ChEBI | CHEBI:37580 |
IUPAC Name | acetyl chloride |
SMILES | CC(Cl)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15415367
|
Thermo Scientific Alfa Aesar
H32616.AE |
100 mL | N/A | ||||||
15425367
|
Thermo Scientific Alfa Aesar
H32616.AP |
500 mL |
£75.78
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-36-5 | |
78.50 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
6367 | |
acetyl chloride |
C2H3ClO | |
MFCD00000719 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CHEBI:37580 | |
CC(Cl)=O |
Specifications
75-36-5 | |
C2H3ClO | |
UN1593 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
1M soln. in dichloromethane |
1.294 | |
MFCD00000719 | |
Moisture sensitive | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.5 | |
Acetyl chloride |
Safety and Handling
GHS H Statement
H351-H302-H315-H319
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H336-H350
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 200-865-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only