Learn More
Acetophenone, 99%, Thermo Scientific Chemicals
£25.93 - £216.08
Chemical Identifiers
CAS | 98-86-2 |
---|---|
Molecular Formula | C8H8O |
Molecular Weight (g/mol) | 120.151 |
MDL Number | MFCD00008724 |
InChI Key | KWOLFJPFCHCOCG-UHFFFAOYSA-N |
Synonym | acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol |
PubChem CID | 7410 |
ChEBI | CHEBI:27632 |
IUPAC Name | 1-phenylethanone |
SMILES | CC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463057
|
Thermo Scientific Alfa Aesar
A12727.AP |
500 mL |
£25.93
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453057
|
Thermo Scientific Alfa Aesar
A12727.0F |
2500 mL |
£76.98
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15443057
|
Thermo Scientific Alfa Aesar
A12727.0D |
10,000 mL |
£216.08
10000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.
Solubility
Miscible with water, chloroform, ethanol, ether, benzene and acetone. Slightly miscible with concentrated sulfuric acid.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Chemical Identifiers
98-86-2 | |
120.151 | |
KWOLFJPFCHCOCG-UHFFFAOYSA-N | |
7410 | |
1-phenylethanone |
C8H8O | |
MFCD00008724 | |
acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol | |
CHEBI:27632 | |
CC(=O)C1=CC=CC=C1 |
Specifications
98-86-2 | |
1.03 | |
76°C (169°F) | |
C6H5COCH3 | |
MFCD00008724 | |
acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol | |
KWOLFJPFCHCOCG-UHFFFAOYSA-N | |
1-phenylethanone | |
7410 | |
120.15 | |
Acetophenone |
19°C to 20°C | |
201°C to 203°C | |
C8H8O | |
1.5325 | |
14,73 | |
Miscible with water,chloroform,ethanol,ether,benzene and acetone. Slightly miscible with concentrated sulfuric acid. | |
CC(=O)C1=CC=CC=C1 | |
120.151 | |
CHEBI:27632 | |
99% |
Safety and Handling
GHS H Statement
H302-H319-H227
Harmful if swallowed.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
H227-H302-H319
missing translation for 'einecsNumber' : 202-708-7
missing translation for 'rtecsNumber' : AM5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Acetophenone, 99%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Acetophenone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.