missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile Anhydrous (DNA Synthesis), Fisher BioReagents

Product Code. 10031693
Click to view available options
Quantity:
450 mL
4 L
Packaging:
Amber Glass
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
Acetonitrile Anhydrous is commonly used in DNA or peptide synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 1 max. at 190nm, 0.15 max. at 195nm, 0.07 max. at 200nm, 0.05 max. at 205nm, 0.04 max. at 210nm, 0.02 max. at 220nm, 0.001 max. at 254nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Boiling Point 81°C
Quantity 450 mL
Packaging Amber Glass
Physical Form Liquid
Refractive Index 1.3405 to 1.3425 (at 25°C)
Assay Percent Range ≥99.9 %
Residue after Evaporation 1.0ppm max.
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
UN Number 1648
Viscosity 0.36 cP at 20°C
Water 10ppm max.
Formula Weight 41.05
Percent Purity ≥99.9%
Grade DNA Synthesis
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'