missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
25.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
25.23 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
25.23 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Packaging | Amber glass bottle |
Quantity | 1 L |
Color | Colorless |
Formula Weight | 58.08g/mol |
Vapor Pressure | 247mbar at 20°C |
Viscosity | 0.32 mPaS at 20°C |
Vapor Density | 2 |
pH | 7 |
Melting Point | -95°C |
Boiling Point | 56°C |
Show More |
Safety and Handling
Product Identifier
- Acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'