missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-2-methylquinoline, 98%, Thermo Scientific Chemicals

£22.50 - £65.00

Chemical Identifiers

CAS 3033-82-7
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD00051764
InChI Key VVLYDFPOGMTMFJ-UHFFFAOYSA-N
Synonym 8-chloroquinaldine, quinaldine, 8-chloro, quinoline, 8-chloro-2-methyl, 8-chloro-2-methyl-quinoline, 2-methyl-8-chloroquinoline, pubchem5852, acmc-1aehk, maybridge1_006766, 8-chloranyl-2-methyl-quinoline, 8-chloro-2-methylquinoline
PubChem CID 221113
IUPAC Name 8-chloro-2-methylquinoline
SMILES CC1=NC2=C(C=CC=C2Cl)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494507
View Documents
Thermo Scientific Alfa Aesar
B20504.03
1 g
£22.50
1g
Please to purchase this item. Need a web account? Register with us today!
11404517
View Documents
Thermo Scientific Alfa Aesar
B20504.06
5 g
£65.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3033-82-7
177.631
VVLYDFPOGMTMFJ-UHFFFAOYSA-N
221113
CC1=NC2=C(C=CC=C2Cl)C=C1
C10H8ClN
MFCD00051764
8-chloroquinaldine, quinaldine, 8-chloro, quinoline, 8-chloro-2-methyl, 8-chloro-2-methyl-quinoline, 2-methyl-8-chloroquinoline, pubchem5852, acmc-1aehk, maybridge1_006766, 8-chloranyl-2-methyl-quinoline, 8-chloro-2-methylquinoline
8-chloro-2-methylquinoline

Specifications

3033-82-7
C10H8ClN
119193
VVLYDFPOGMTMFJ-UHFFFAOYSA-N
8-chloro-2-methylquinoline
221113
98%
65°C to 68°C
MFCD00051764
8-chloroquinaldine, quinaldine, 8-chloro, quinoline, 8-chloro-2-methyl, 8-chloro-2-methyl-quinoline, 2-methyl-8-chloroquinoline, pubchem5852, acmc-1aehk, maybridge1_006766, 8-chloranyl-2-methyl-quinoline, 8-chloro-2-methylquinoline
CC1=NC2=C(C=CC=C2Cl)C=C1
177.631
177.63
8-Chloro-2-methylquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only