Learn More
7-Bromoheptanenitrile, 98%, Thermo Scientific Chemicals
£46.03 - £193.97
Chemical Identifiers
CAS | 20965-27-9 |
---|---|
Molecular Formula | C7H12BrN |
Molecular Weight (g/mol) | 190.08 |
MDL Number | MFCD00001982 |
InChI Key | HVVQSKCGHAPHMV-UHFFFAOYSA-N |
Synonym | 7-bromoheptanonitrile, heptanenitrile, 7-bromo, 6-cyanohexyl bromide, 1-bromo-6-cyanohexane, 6-bromohexylcyanide, 6-bromohexyl cyanide, 7-bromo-heptanonitrile, 7-bromoheptan enitrile, 7-bromanylheptanenitrile, acmc-1cc07 |
PubChem CID | 88737 |
IUPAC Name | 7-bromoheptanenitrile |
SMILES | BrCCCCCCC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341817
|
Thermo Scientific Alfa Aesar
L05068.06 |
5 g |
£46.03
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351817
|
Thermo Scientific Alfa Aesar
L05068.14 |
25 g |
£193.97
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20965-27-9 | |
190.08 | |
HVVQSKCGHAPHMV-UHFFFAOYSA-N | |
88737 | |
BrCCCCCCC#N |
C7H12BrN | |
MFCD00001982 | |
7-bromoheptanonitrile, heptanenitrile, 7-bromo, 6-cyanohexyl bromide, 1-bromo-6-cyanohexane, 6-bromohexylcyanide, 6-bromohexyl cyanide, 7-bromo-heptanonitrile, 7-bromoheptan enitrile, 7-bromanylheptanenitrile, acmc-1cc07 | |
7-bromoheptanenitrile |
Specifications
20965-27-9 | |
114°C to 115°C (4 mmHg) | |
C7H12BrN | |
MFCD00001982 | |
1748070 | |
HVVQSKCGHAPHMV-UHFFFAOYSA-N | |
7-bromoheptanenitrile | |
88737 | |
98% |
1.266 | |
>110°C (230°F) | |
1.475 | |
UN3276 | |
7-bromoheptanonitrile, heptanenitrile, 7-bromo, 6-cyanohexyl bromide, 1-bromo-6-cyanohexane, 6-bromohexylcyanide, 6-bromohexyl cyanide, 7-bromo-heptanonitrile, 7-bromoheptan enitrile, 7-bromanylheptanenitrile, acmc-1cc07 | |
BrCCCCCCC#N | |
190.08 | |
190.09 | |
7-Bromoheptanenitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 244-128-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only