missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinecarbaldehyde, 97%, Thermo Scientific™

£17.30 - £163.00

Chemical Identifiers

CAS 4113-04-6
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
InChI Key VUAOIXANWIFYCU-UHFFFAOYSA-N
Synonym quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
PubChem CID 765653
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10055712
SDS View Documents
Thermo Scientific Maybridge
CC04504CB
250 mg Amber Glass Bottle
£17.30
250mg
Please to purchase this item. Need a web account? Register with us today!
10164262
SDS View Documents
Thermo Scientific Maybridge
CC04504DA
1 g Amber Glass Bottle
£51.80
1g
Please to purchase this item. Need a web account? Register with us today!
10021384
SDS View Documents
Thermo Scientific Maybridge
CC04504DE
5 g Amber Glass Bottle
£163.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

4113-04-6
157.17
quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
C10H7NO
VUAOIXANWIFYCU-UHFFFAOYSA-N
765653

Specifications

4113-04-6
C10H7NO
VUAOIXANWIFYCU-UHFFFAOYSA-N
765653
97%
97%
quinoline-6-carbaldehyde, 6-quinolinecarboxaldehyde, 6-quinolinecarbaldehyde, quinoline-6-carboxaldehyde, 6-formylquinoline, 6-quinolinemethanal, pubchem10181, quinolin-6-carbaldehyde, acmc-2097el
157.17
157.17
6-Quinolinecarbaldehyde
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.