missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholinonicotinic acid, 95+%, Thermo Scientific™

£31.10 - £137.00

Chemical Identifiers

CAS 120800-52-4
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.217
InChI Key XXDSDFLDYNISKD-UHFFFAOYSA-N
Synonym 6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl
PubChem CID 2776460
IUPAC Name 6-morpholin-4-ylpyridine-3-carboxylic acid
SMILES C1COCCN1C2=NC=C(C=C2)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10161112
View Documents
Thermo Scientific Maybridge
CC17501CB
250 mg Amber Glass Bottle
£31.10
250mg
Please to purchase this item. Need a web account? Register with us today!
10732103
View Documents
Thermo Scientific Maybridge
CC17501DA
1 g Amber Glass Bottle
£42.60
1g
Please to purchase this item. Need a web account? Register with us today!
10081404
View Documents
Thermo Scientific Maybridge
CC17501DE
5 g Amber Glass Bottle
£137.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

120800-52-4
208.217
6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl
6-morpholin-4-ylpyridine-3-carboxylic acid
C10H12N2O3
XXDSDFLDYNISKD-UHFFFAOYSA-N
2776460
C1COCCN1C2=NC=C(C=C2)C(=O)O

Specifications

120800-52-4
C10H12N2O3
XXDSDFLDYNISKD-UHFFFAOYSA-N
6-morpholin-4-ylpyridine-3-carboxylic acid
2776460
95%
95%
6-morpholinonicotinic acid, 6-morpholin-4-ylnicotinic acid, 6-morpholin-4-yl pyridine-3-carboxylic acid, 6-4-morpholinyl-3-pyridinecarboxylic acid, 6-morpholin-4-yl-nicotinic acid, 3-pyridinecarboxylic acid, 6-4-morpholinyl, 6-morpholinopyridine-3-carboxylic acid, pubchem19467, acmc-209a8e, 3-pyridinecarboxylicacid, 6-4-morpholinyl
C1COCCN1C2=NC=C(C=C2)C(=O)O
208.217
208.22
6-Morpholinonicotinic acid
Videos
SDS
Documents

Documents

Product Certifications