missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, Thermo Scientific™
£29.90 - £401.00
Chemical Identifiers
CAS | 1192758-40-9 |
---|---|
Molecular Formula | C9H14ClN3O4 |
Molecular Weight (g/mol) | 263.678 |
InChI Key | VARLCSQEQQKXKH-UHFFFAOYSA-N |
Synonym | 6-morpholinopyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-yl pyridazine-3-carboxylic acid hydrate hydrochloride |
PubChem CID | 53444630 |
IUPAC Name | 6-morpholin-4-ylpyridazine-3-carboxylic acid;hydrate;hydrochloride |
SMILES | C1COCCN1C2=NN=C(C=C2)C(=O)O.O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10753717
|
Thermo Scientific Maybridge
CC77751CB |
250 mg | Amber Glass Bottle |
£29.90
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10046724
|
Thermo Scientific Maybridge
CC77751DA |
1 g | Amber Glass Bottle |
£98.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10650096
|
Thermo Scientific Maybridge
CC77751DE |
5 g | Amber Glass Bottle |
£401.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1192758-40-9 | |
263.678 | |
6-morpholinopyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-yl pyridazine-3-carboxylic acid hydrate hydrochloride | |
6-morpholin-4-ylpyridazine-3-carboxylic acid;hydrate;hydrochloride |
C9H14ClN3O4 | |
VARLCSQEQQKXKH-UHFFFAOYSA-N | |
53444630 | |
C1COCCN1C2=NN=C(C=C2)C(=O)O.O.Cl |
Specifications
1192758-40-9 | |
6-morpholinopyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-yl pyridazine-3-carboxylic acid hydrate hydrochloride | |
C1COCCN1C2=NN=C(C=C2)C(=O)O.O.Cl | |
263.678 | |
245.67 | |
6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate |
C9H14ClN3O4 | |
VARLCSQEQQKXKH-UHFFFAOYSA-N | |
6-morpholin-4-ylpyridazine-3-carboxylic acid;hydrate;hydrochloride | |
53444630 | |
97% |