missing translation for 'onlineSavingsMsg'
Learn More

6-Methyl-3,5-heptadien-2-one, 97%, Thermo Scientific Chemicals

£26.03 - £78.70

Chemical Identifiers

CAS 1604-28-0
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00043647
InChI Key KSKXSFZGARKWOW-GQCTYLIASA-N
Synonym 6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
PubChem CID 5370101
IUPAC Name (3E)-6-methylhepta-3,5-dien-2-one
SMILES CC(=CC=CC(=O)C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377256
View Documents
Thermo Scientific Alfa Aesar
L09169.06
5 g
£26.03
5g
Please to purchase this item. Need a web account? Register with us today!
11387256
View Documents
Thermo Scientific Alfa Aesar
L09169.14
25 g
£78.70
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

1604-28-0
124.183
KSKXSFZGARKWOW-GQCTYLIASA-N
5370101
CC(=CC=CC(=O)C)C
C8H12O
MFCD00043647
6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
(3E)-6-methylhepta-3,5-dien-2-one

Specifications

1604-28-0
95°C to 96°C (20 mmHg)
C8H12O
MFCD00043647
6-methyl-3,5-heptadien-2-one, 6-methylhepta-3,5-dien-2-one, methylheptadienone, unii-p7cmp2e76c, 3,5-heptadien-2-one, 6-methyl, 3e-6-methylhepta-3,5-dien-2-one, 3,5-heptadien-2-one, 6-methyl-, e, 6-methyl-trans-3,5-heptadien-2-one, p7cmp2e76c, fema no. 3363
KSKXSFZGARKWOW-GQCTYLIASA-N
(3E)-6-methylhepta-3,5-dien-2-one
5370101
97%
0.89
Pleasant
1.534
1699937
Miscible with alcohol. Immiscible with water.
CC(=CC=CC(=O)C)C
124.183
124.18
6-Methyl-3,5-heptadien-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'einecsNumber' : 216-507-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 6-Methyl-3,5-heptadien-2-one

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 6-methyl-3,5-heptadien-2-one

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.