missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinoyl chloride, 98%, Thermo Scientific Chemicals

£26.50 - £221.00

Chemical Identifiers

CAS 58757-38-3
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 175.996
MDL Number MFCD00051775
InChI Key FMEBIWNKYZUWFV-UHFFFAOYSA-N
Synonym 6-chloronicotinoyl chloride, 6-chloronicotinoylchloride, 2-chloropyridine-5-carbonyl chloride, 3-pyridinecarbonylchloride, 6-chloro, 3-pyridinecarbonyl chloride, 6-chloro, 6-chloro-nicotinoyl chloride, pubchem18665, 6-chloronicotinic chloride, acmc-1ay8e, 6-chloro-nicotinic chloride
PubChem CID 2782165
IUPAC Name 6-chloropyridine-3-carbonyl chloride
SMILES C1=CC(=NC=C1C(=O)Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459483
SDS View Documents
Thermo Scientific Alfa Aesar
A12354.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11469483
SDS View Documents
Thermo Scientific Alfa Aesar
A12354.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11479483
SDS View Documents
Thermo Scientific Alfa Aesar
A12354.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58757-38-3
175.996
FMEBIWNKYZUWFV-UHFFFAOYSA-N
2782165
C1=CC(=NC=C1C(=O)Cl)Cl
C6H3Cl2NO
MFCD00051775
6-chloronicotinoyl chloride, 6-chloronicotinoylchloride, 2-chloropyridine-5-carbonyl chloride, 3-pyridinecarbonylchloride, 6-chloro, 3-pyridinecarbonyl chloride, 6-chloro, 6-chloro-nicotinoyl chloride, pubchem18665, 6-chloronicotinic chloride, acmc-1ay8e, 6-chloro-nicotinic chloride
6-chloropyridine-3-carbonyl chloride

Specifications

58757-38-3
84°C to 85°C (3 mmHg)
MFCD00051775
115927
FMEBIWNKYZUWFV-UHFFFAOYSA-N
6-chloropyridine-3-carbonyl chloride
2782165
98%
48°C to 52°C
C6H3Cl2NO
UN3261
6-chloronicotinoyl chloride, 6-chloronicotinoylchloride, 2-chloropyridine-5-carbonyl chloride, 3-pyridinecarbonylchloride, 6-chloro, 3-pyridinecarbonyl chloride, 6-chloro, 6-chloro-nicotinoyl chloride, pubchem18665, 6-chloronicotinic chloride, acmc-1ay8e, 6-chloro-nicotinic chloride
C1=CC(=NC=C1C(=O)Cl)Cl
175.996
176
6-Chloronicotinoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.