missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2-benzothiazolinone, 98%, Thermo Scientific™

£34.80 - £138.00

Chemical Identifiers

CAS 62266-82-4
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.08
MDL Number MFCD00239363
InChI Key HECJMTPEVWQFCY-UHFFFAOYSA-N
Synonym 6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
PubChem CID 188444
SMILES BrC1=CC=C2NC(=O)SC2=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15456848
View Documents
Thermo Scientific Alfa Aesar
H60385.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15466848
View Documents
Thermo Scientific Alfa Aesar
H60385.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

62266-82-4
230.08
HECJMTPEVWQFCY-UHFFFAOYSA-N
188444
C7H4BrNOS
MFCD00239363
6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
BrC1=CC=C2NC(=O)SC2=C1

Specifications

62266-82-4
C7H4BrNOS
6-bromo-2-benzothiazolinone, 6-bromobenzo d thiazol-2 3h-one, 6-bromo-2 3h-benzothiazolone, 6-bromo-3h-benzothiazol-2-one, 2 3h-benzothiazolone, 6-bromo, 6-bromo-1,3-benzothiazol-2 3h-one, 6-bromo-2-hydroxy benzothiazol, 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one, 2-benzothiazolol, 6-bromo, pubchem21839
BrC1=CC=C2NC(=O)SC2=C1
230.08
230.08
6-Bromo-2-benzothiazolinone
229°C to 233°C
MFCD00239363
HECJMTPEVWQFCY-UHFFFAOYSA-N
6-bromo-3H-1,3-benzothiazol-2-one
188444
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates