Learn More
6-Amino-1-hexanol, 97%, Thermo Scientific Chemicals
£58.29
Chemical Identifiers
CAS | 4048-33-3 |
---|---|
Molecular Formula | C6H15NO |
Molecular Weight (g/mol) | 117.192 |
MDL Number | MFCD00008241 |
InChI Key | SUTWPJHCRAITLU-UHFFFAOYSA-N |
Synonym | 6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol |
PubChem CID | 19960 |
IUPAC Name | 6-aminohexan-1-ol |
SMILES | C(CCCO)CCN |
Description
6-Amino-1-hexanol is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Amino-1-hexanol is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture.
Chemical Identifiers
4048-33-3 | |
117.192 | |
SUTWPJHCRAITLU-UHFFFAOYSA-N | |
19960 | |
C(CCCO)CCN |
C6H15NO | |
MFCD00008241 | |
6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol | |
6-aminohexan-1-ol |
Specifications
4048-33-3 | |
136°C to 139°C (30 mmHg) | |
C6H15NO | |
1732524 | |
6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol | |
SUTWPJHCRAITLU-UHFFFAOYSA-N | |
6-aminohexan-1-ol | |
19960 | |
97% |
55°C to 58°C | |
133°C (271°F) | |
MFCD00008241 | |
Hygroscopic | |
Soluble in water. | |
C(CCCO)CCN | |
117.192 | |
117.19 | |
6-Amino-1-hexanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 223-748-1
missing translation for 'rtecsNumber' : MO8840000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only