Learn More
Greiner Bio-One 5 mL Sterile Serological Pipets
GREENER_CHOICE
Drip-free pipetting design with high optical clarity and maximum accuracy.
£50.61 - £91.11
Specifications
Material Polystyrene
Sterility Sterile
Graduated Yes
Graduations 1/10
Volume (Metric) 5 mL
View More Specs
Products 2
Product Code Brand Packaging Type Price Quantity & Availability  
Product Code Brand Packaging Type Price Quantity & Availability  
13002013
GREENER_CHOICE
View Documents
Greiner Bio-One
606160
Individual Plastic Wrap
£50.61
Pack of 200
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
10018810
GREENER_CHOICE
View Documents
Greiner Bio-One
606107
Bulk
£91.11
Pack of 500
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
Description
Description
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
Specifications
Specifications
Polystyrene
Yes
5 mL
Sterile
1/10
Serological Pipet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Greiner Bio-One 5 mL Sterile Serological Pipets