missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole, 99%, Thermo Scientific Chemicals

£37.39 - £695.46

Chemical Identifiers

CAS 399-52-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00005671
InChI Key ODFFPRGJZRXNHZ-UHFFFAOYSA-N
Synonym 5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
PubChem CID 67861
ChEBI CHEBI:72818
IUPAC Name 5-fluoro-1H-indole
SMILES FC1=CC=C2NC=CC2=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11428266
SDS View Documents
Thermo Scientific Alfa Aesar
A15346.03
1 g
£37.39
1g
Please to purchase this item. Need a web account? Register with us today!
11438266
SDS View Documents
Thermo Scientific Alfa Aesar
A15346.06
5 g
£164.82
5g
Please to purchase this item. Need a web account? Register with us today!
11448266
SDS View Documents
Thermo Scientific Alfa Aesar
A15346.14
25 g
£695.46
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

399-52-0
135.14
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
67861
5-fluoro-1H-indole
C8H6FN
MFCD00005671
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
CHEBI:72818
FC1=CC=C2NC=CC2=C1

Specifications

399-52-0
>110°C (230°F)
MFCD00005671
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
FC1=CC=C2NC=CC2=C1
135.14
CHEBI:72818
99%
42°C to 47°C
C8H6FN
112350
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
5-fluoro-1H-indole
67861
135.14
5-Fluoroindole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 206-917-4

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.