missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxy-3-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

£59.50 - £206.00

Chemical Identifiers

CAS 23602-63-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00275687
InChI Key NSKZAOKQZDLHGO-UHFFFAOYSA-N
Synonym benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
PubChem CID 2774358
IUPAC Name 5-chloro-2-hydroxy-3-methylbenzaldehyde
SMILES CC1=C(C(=CC(=C1)Cl)C=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339755
View Documents
Thermo Scientific Alfa Aesar
L19793.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11349755
View Documents
Thermo Scientific Alfa Aesar
L19793.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

23602-63-3
170.592
NSKZAOKQZDLHGO-UHFFFAOYSA-N
2774358
CC1=C(C(=CC(=C1)Cl)C=O)O
C8H7ClO2
MFCD00275687
benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
5-chloro-2-hydroxy-3-methylbenzaldehyde

Specifications

23602-63-3
C8H7ClO2
2249708
benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
CC1=C(C(=CC(=C1)Cl)C=O)O
170.592
170.6
5-Chloro-2-hydroxy-3-methylbenzaldehyde
59°C
MFCD00275687
Air Sensitive
NSKZAOKQZDLHGO-UHFFFAOYSA-N
5-chloro-2-hydroxy-3-methylbenzaldehyde
2774358
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.