missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4-dichloropyrimidine, 97%, Thermo Scientific Chemicals

£14.50 - £135.00

Chemical Identifiers

CAS 5177-27-5
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.989
MDL Number MFCD05662684
InChI Key RINHVELYMZLXIW-UHFFFAOYSA-N
PubChem CID 257797
IUPAC Name 2,4-dichloropyrimidin-5-amine
SMILES C1=C(C(=NC(=N1)Cl)Cl)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15435357
View Documents
Thermo Scientific Alfa Aesar
H32606.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15415357
View Documents
Thermo Scientific Alfa Aesar
H32606.03
1 g
£40.60
1g
Please to purchase this item. Need a web account? Register with us today!
15425357
View Documents
Thermo Scientific Alfa Aesar
H32606.06
5 g
£135.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5177-27-5
163.989
RINHVELYMZLXIW-UHFFFAOYSA-N
2,4-dichloropyrimidin-5-amine
C4H3Cl2N3
MFCD05662684
257797
C1=C(C(=NC(=N1)Cl)Cl)N

Specifications

5177-27-5
MFCD05662684
RINHVELYMZLXIW-UHFFFAOYSA-N
2,4-dichloropyrimidin-5-amine
257797
97%
C4H3Cl2N3
5-amino-2,4-dichloropyrimidine, 2,4-dichloro-5-aminopyrimidine, 2,4-dichloro-pyrimidin-5-ylamine, 2,4-dichloro-5-pyrimidinamine, 5-amino-2, 4-dichloropyrimidine, 5-pyrimidinamine, 2,4-dichloro, 2,4-dichlorpyrimidin-5-amin, pubchem21459, acmc-209kvs, ksc497e8n
C1=C(C(=NC(=N1)Cl)Cl)N
163.989
164
5-Amino-2,4-dichloropyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only