missing translation for 'onlineSavingsMsg'
Learn More

5,6,7-Trimethoxyflavone, 97%, Thermo Scientific™

Product Code. 11495297
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 973-67-1
Molecular Formula C18H16O5
Molecular Weight (g/mol) 312.32
MDL Number MFCD00017457
InChI Key HJNJAUYFFFOFBW-UHFFFAOYSA-N
Synonym 5,6,7-trimethoxyflavone, baicalein trimethyl ether, baicalein 5,6,7-trimethyl ether, baicalein-5,6,7-trimethylether, 5,6,7-trimethoxy-2-phenyl-4h-chromen-4-one, 5,6,7-trimethoxy-2-phenyl-chromen-4-one, baicalein-5,6,7-trimethyl ether, pubchem9835, 5,6,7-trimethylbaicalein
PubChem CID 442583
ChEBI CHEBI:2980
IUPAC Name 5,6,7-trimethoxy-2-phenylchromen-4-one
SMILES COC1=C(OC)C(OC)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1

Specifications

Melting Point 165°C to 167°C
Odor Odorless
Quantity 0.25 g
Formula Weight 312.33
Percent Purity 97%
Chemical Name or Material 5,6,7-Trimethoxyflavone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'