missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Dimethylaminobenzylidene)rhodanine 98%, Thermo Scientific Chemicals

£33.17 - £98.29

Chemical Identifiers

CAS 536-17-4
Molecular Formula C12H12N2OS2
Molecular Weight (g/mol) 264.361
MDL Number MFCD00064857
InChI Key JJRVRELEASDUMY-UHFFFAOYSA-N
Synonym unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
PubChem CID 2723826
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493367
View Documents
Thermo Scientific Alfa Aesar
A17152.06
5 g
£33.17
5g
Please to purchase this item. Need a web account? Register with us today!
11413377
View Documents
Thermo Scientific Alfa Aesar
A17152.14
25 g
£98.29
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.

Solubility
Insoluble in water, 993.7(mg/L) at 25°C

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications

Chemical Identifiers

536-17-4
264.361
JJRVRELEASDUMY-UHFFFAOYSA-N
2723826
CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2
C12H12N2OS2
MFCD00064857
unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one

Specifications

536-17-4
Weak
MFCD00064857
14,3231
Insoluble in water,993.7(mg/L) at 25°C
CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2
264.361
264.37
5-(4-Dimethylaminobenzylidene)rhodanine 98%
275°C to 280°C
C12H12N2OS2
189065
unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
JJRVRELEASDUMY-UHFFFAOYSA-N
5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
2723826
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-625-2

missing translation for 'rtecsNumber' : VI8090000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.