missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylglyoxal hydrate, Thermo Scientific™

Product Code. 10537390
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 1736-56-7
Packaging Amber Glass Bottle
Molecular Formula C9H5F3O2
Quantity 1 g
Synonym 2-oxo-2-4-trifluoromethyl phenyl acetaldehyde, 2-oxo-2-4-trifluoromethyl phenyl ethanal, 2-oxidanylidene-2-4-trifluoromethyl phenyl ethanal, 4-trifluoromethylphenyl oxo acetaldehyde, oxo 4-trifluoromethyl phenyl acetaldehyde, 1-4-trifluoromethyl phenyl-1,2-ethanedione
InChI Key BGOMXTCPIUNFKR-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=O)C=O)C(F)(F)F
IUPAC Name 2-oxo-2-[4-(trifluoromethyl)phenyl]acetaldehyde
Molecular Weight (g/mol) 202.132
PubChem CID 2783286
Formula Weight 202.13
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)phenylglyoxal hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.