missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylaniline, 95%, Thermo Scientific™

£101.00 - £250.00

Chemical Identifiers

CAS 69491-57-2
Molecular Formula C10H9N3
Molecular Weight (g/mol) 171.203
MDL Number MFCD09817490
InChI Key RUVNVJCKWOOLOH-UHFFFAOYSA-N
PubChem CID 12456170
IUPAC Name 4-pyrimidin-2-ylaniline
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10245654
SDS View Documents
Thermo Scientific Maybridge
CC56214CB
250 mg Amber Glass Bottle
£101.00
250mg
Please to purchase this item. Need a web account? Register with us today!
10682855
SDS View Documents
Thermo Scientific Maybridge
CC56214DA
1 g Amber Glass Bottle
£250.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

69491-57-2
171.203
RUVNVJCKWOOLOH-UHFFFAOYSA-N
4-pyrimidin-2-ylaniline
C10H9N3
MFCD09817490
12456170
C1=CN=C(N=C1)C2=CC=C(C=C2)N

Specifications

69491-57-2
100.0
C10H9N3
4-pyrimidin-2-yl aniline, 2-4-aminophenyl pyrimidine, p-aminophenylpyrimidine, 4-2-pyrimidinyl aniline, 4-pyrimidin-2-ylphenylamine, benzenamine,4-2-pyrimidinyl, benzenamine, 4-2-pyrimidinyl
C1=CN=C(N=C1)C2=CC=C(C=C2)N
171.203
171.2
4-Pyrimidin-2-ylaniline
96.5
95%
MFCD09817490
RUVNVJCKWOOLOH-UHFFFAOYSA-N
4-pyrimidin-2-ylaniline
12456170
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.