missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid, 97%, Thermo Scientific™

£20.70 - £292.00

Chemical Identifiers

CAS 56671-28-4
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00052170
InChI Key FABBWECRHZNMDQ-UHFFFAOYSA-N
Synonym 4-oxo-4,5,6,7-tetrahydrobenzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydrobenzo b furan-3-carboxylic acid, maybridge1_007745, 4-oxo-5,6,7-trihydrobenzo 1,2-b furan-3-carboxylic acid, oxotetrahydrobenzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo-3-benzofurancarboxylic acid, 4-oxo-4,5,6,7-tetrahydrocoumarone-3-carboxylic acid, 3-benzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo, 4,5,6,7-tetrahydro-4-oxobenzofuran-3-carboxylic acid
PubChem CID 713937
IUPAC Name 4-oxo-6,7-dihydro-5H-1-benzofuran-3-carboxylic acid
SMILES C1CC2=C(C(=O)C1)C(=CO2)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10630923
SDS View Documents
Thermo Scientific Maybridge
SEW02412CB
250 mg
£20.70
250mg
Please to purchase this item. Need a web account? Register with us today!
10783633
SDS View Documents
Thermo Scientific Maybridge
SEW02412DA
1 g
£55.20
1g
Please to purchase this item. Need a web account? Register with us today!
10546524
SDS View Documents
Thermo Scientific Maybridge
SEW02412EA
10 g
£292.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

56671-28-4
180.159
FABBWECRHZNMDQ-UHFFFAOYSA-N
713937
C1CC2=C(C(=O)C1)C(=CO2)C(=O)O
C9H8O4
MFCD00052170
4-oxo-4,5,6,7-tetrahydrobenzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydrobenzo b furan-3-carboxylic acid, maybridge1_007745, 4-oxo-5,6,7-trihydrobenzo 1,2-b furan-3-carboxylic acid, oxotetrahydrobenzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo-3-benzofurancarboxylic acid, 4-oxo-4,5,6,7-tetrahydrocoumarone-3-carboxylic acid, 3-benzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo, 4,5,6,7-tetrahydro-4-oxobenzofuran-3-carboxylic acid
4-oxo-6,7-dihydro-5H-1-benzofuran-3-carboxylic acid

Specifications

56671-28-4
97%
C9H8O4
4-oxo-4,5,6,7-tetrahydrobenzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydrobenzo b furan-3-carboxylic acid, maybridge1_007745, 4-oxo-5,6,7-trihydrobenzo 1,2-b furan-3-carboxylic acid, oxotetrahydrobenzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo-3-benzofurancarboxylic acid, 4-oxo-4,5,6,7-tetrahydrocoumarone-3-carboxylic acid, 3-benzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo, 4,5,6,7-tetrahydro-4-oxobenzofuran-3-carboxylic acid
C1CC2=C(C(=O)C1)C(=CO2)C(=O)O
180.159
180.16
4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid
97.0
Amber Glass Bottle
MFCD00052170
FABBWECRHZNMDQ-UHFFFAOYSA-N
4-oxo-6,7-dihydro-5H-1-benzofuran-3-carboxylic acid
713937
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.