missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-oxo-4-(2-thienyl)butanoic acid, Thermo Scientific™
£8.10 - £54.10
Chemical Identifiers
CAS | 4653-08-1 |
---|---|
Molecular Formula | C8H8O3S |
Molecular Weight (g/mol) | 184.21 |
InChI Key | ULJMYWHLMLRYSO-UHFFFAOYSA-N |
Synonym | 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid |
PubChem CID | 78385 |
ChEBI | CHEBI:64434 |
IUPAC Name | 4-oxo-4-thiophen-2-ylbutanoic acid |
SMILES | C1=CSC(=C1)C(=O)CCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10279962
|
Thermo Scientific Maybridge
BTB10644DA |
1 g |
£8.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10749203
|
Thermo Scientific Maybridge
BTB10644EA |
10 g |
£27.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10774277
|
Thermo Scientific Maybridge
BTB10644EB |
25 g |
£54.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
4653-08-1 | |
184.21 | |
3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid | |
CHEBI:64434 | |
C1=CSC(=C1)C(=O)CCC(=O)O |
C8H8O3S | |
ULJMYWHLMLRYSO-UHFFFAOYSA-N | |
78385 | |
4-oxo-4-thiophen-2-ylbutanoic acid |
Specifications
4653-08-1 | |
C8H8O3S | |
ULJMYWHLMLRYSO-UHFFFAOYSA-N | |
4-oxo-4-thiophen-2-ylbutanoic acid | |
78385 | |
184.21 | |
4-Oxo-4-(2-thienyl)butanoic acid |
97% | |
3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid | |
C1=CSC(=C1)C(=O)CCC(=O)O | |
184.21 | |
CHEBI:64434 | |
97% |
Safety and Handling
GHS Signal Word: Warning
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title