Learn More
4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate, 95%, Thermo Scientific Chemicals
£42.76 - £122.61
Chemical Identifiers
CAS | 80149-80-0 |
---|---|
Molecular Formula | C12H17NO5Si |
Molecular Weight (g/mol) | 283.36 |
MDL Number | MFCD00042930 |
InChI Key | ZAQWGGKIMQIVGM-UHFFFAOYSA-N |
Synonym | 4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204 |
PubChem CID | 3086116 |
IUPAC Name | (4-nitrophenyl) 2-trimethylsilylethyl carbonate |
SMILES | C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435776
|
Thermo Scientific Alfa Aesar
H60466.03 |
1 g |
£42.76
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445776
|
Thermo Scientific Alfa Aesar
H60466.06 |
5 g |
£122.61
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as a reagent for the introduction of the TEOC-amino protecting group which can be cleaved by F-.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a reagent for the introduction of the TEOC-amino protecting group which can be cleaved by F-.
Solubility
Slightly soluble in water.
Notes
Moisture & Light Sensitive. Store at 2-8°C.
Chemical Identifiers
80149-80-0 | |
283.36 | |
ZAQWGGKIMQIVGM-UHFFFAOYSA-N | |
3086116 | |
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O |
C12H17NO5Si | |
MFCD00042930 | |
4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204 | |
(4-nitrophenyl) 2-trimethylsilylethyl carbonate |
Specifications
80149-80-0 | |
C12H17NO5Si | |
MFCD00042930 | |
Light and moisture sensitive | |
Slightly soluble in water. | |
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+]([O-])=O | |
283.36 | |
283.35 | |
4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate |
35°C to 40°C | |
C12H17NO5Si | |
4320585 | |
4-nitrophenyl 2-trimethylsilyl ethyl carbonate, teoc-onp, 2-trimethylsilyl ethyl p-nitrophenyl carbonate, teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate, 4-nitrophenyl 2-trimethylsilylethyl carbonate, 2-trimethylsilyl ethyl 4-nitrophenyl carbonate, 4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene, carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester, ambotzrl-8204 | |
ZAQWGGKIMQIVGM-UHFFFAOYSA-N | |
(4-nitrophenyl) 2-trimethylsilylethyl carbonate | |
3086116 | |
95% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 279-406-7
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only