missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, 99%, Thermo Scientific Chemicals

£34.37 - £255.27

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11466967
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14376.30
250 g
£34.37
250g
Please to purchase this item. Need a web account? Register with us today!
11456967
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14376.0B
1000 g
£68.34
1000g
Please to purchase this item. Need a web account? Register with us today!
11476967
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14376.0I
5000 g
£255.27
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

Solubility
Freely soluble in alcohol, chloroform, ether. Freely soluble in acetone. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

100-02-7
139.11
BTJIUGUIPKRLHP-UHFFFAOYSA-N
980
4-nitrophenol
C6H5NO3
MFCD00007331
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
CHEBI:16836
OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

100-02-7
1.48
169°C (336°F)
MFCD00007331
1281877
14,6621
Freely soluble in alcohol,chloroform,ether; Freely soluble in acetone; Slightly soluble in water.
OC1=CC=C(C=C1)[N+]([O-])=O
139.11
CHEBI:16836
99%
110°C to 115°C
279°C (decomposition)
C6H5NO3
UN1663
Light sensitive
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
BTJIUGUIPKRLHP-UHFFFAOYSA-N
4-nitrophenol
980
139.11
4-Nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H373-H302-H312-H332
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P312-P314-P330-P363-P501c

H301-H312+H332-H373-H500

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS

missing translation for 'einecsNumber' : 202-811-7

missing translation for 'rtecsNumber' : SM2275000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 4-Nitrophenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contains : p-Nitrophenol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only