missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxyphenol, 98%, Thermo Scientific™

£142.00 - £407.00

Chemical Identifiers

CAS 18979-55-0
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00002337
InChI Key XIIIHRLCKLSYNH-UHFFFAOYSA-N
Synonym 4-hexyloxyphenol, 4-hexyloxy phenol, phenol, 4-hexyloxy, 4-n-hexyloxyphenol, p-hexyloxyphenol, unii-35ims9l1fe, 35ims9l1fe, pubchem13208, acmc-1c6hg, hydroquinone monohexyl ether
PubChem CID 29354
ChEBI CHEBI:34407
IUPAC Name 4-hexoxyphenol
SMILES CCCCCCOC1=CC=C(O)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469223
View Documents
Thermo Scientific Alfa Aesar
A11711.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11479223
View Documents
Thermo Scientific Alfa Aesar
A11711.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

18979-55-0
194.27
XIIIHRLCKLSYNH-UHFFFAOYSA-N
29354
4-hexoxyphenol
C12H18O2
MFCD00002337
4-hexyloxyphenol, 4-hexyloxy phenol, phenol, 4-hexyloxy, 4-n-hexyloxyphenol, p-hexyloxyphenol, unii-35ims9l1fe, 35ims9l1fe, pubchem13208, acmc-1c6hg, hydroquinone monohexyl ether
CHEBI:34407
CCCCCCOC1=CC=C(O)C=C1

Specifications

18979-55-0
C12H18O2
473079
XIIIHRLCKLSYNH-UHFFFAOYSA-N
4-hexoxyphenol
29354
194.28
4-n-Hexyloxyphenol
44°C to 47°C
MFCD00002337
4-hexyloxyphenol, 4-hexyloxy phenol, phenol, 4-hexyloxy, 4-n-hexyloxyphenol, p-hexyloxyphenol, unii-35ims9l1fe, 35ims9l1fe, pubchem13208, acmc-1c6hg, hydroquinone monohexyl ether
CCCCCCOC1=CC=C(O)C=C1
194.27
CHEBI:34407
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 242-713-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications