missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxybenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11441003
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39649-71-3
Molecular Formula C13H17ClO2
Molecular Weight (g/mol) 240.727
MDL Number MFCD00000690
InChI Key DQQOONVCLQZWOY-UHFFFAOYSA-N
PubChem CID 170235
IUPAC Name 4-hexoxybenzoyl chloride
SMILES CCCCCCOC1=CC=C(C=C1)C(=O)Cl

Specifications

Boiling Point 203°C to 205°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3265
Beilstein 2210619
Synonym 4-n-hexyloxybenzoyl chloride, 4-hexyloxybenzoyl chloride, 4-hexyloxy benzoyl chloride, benzoyl chloride, 4-hexyloxy, p-hexyloxybenzoyl chloride, 4-hexyloxybenzoylchloride, 4-hexyloxy-benzoyl chloride, 4-hexyloxybenzoic acid chloride, 4-n-hexyl-oxybenzoyl chloride, benzoyl chloride,4-hexyloxy
Formula Weight 240.73
Percent Purity 98%
Chemical Name or Material 4-n-Hexyloxybenzoyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.