missing translation for 'onlineSavingsMsg'
Learn More

4-isocyanato-4-phenyltetrahydropyran, 97%, Thermo Scientific™

£80.50 - £197.00

Chemical Identifiers

CAS 941717-02-8
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.24
MDL Number MFCD11506354
InChI Key HKKGHFQAYVPNAN-UHFFFAOYSA-N
Synonym 4-isocyanato-4-phenyltetrahydropyran, 4-isocyanato-4-phenyltetrahydro-2h-pyran, 4-phenyltetrahydro-2h-pyran-4-yl isocyanate, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate
PubChem CID 33589534
IUPAC Name 4-isocyanato-4-phenyloxane
SMILES O=C=NC1(CCOCC1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10235764
SDS View Documents
Thermo Scientific Maybridge
CC69106CB
250 mg Amber Glass Bottle
£80.50
250mg
Please to purchase this item. Need a web account? Register with us today!
10472824
SDS View Documents
Thermo Scientific Maybridge
CC69106DA
1 g Amber Glass Bottle
£197.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

941717-02-8
203.24
HKKGHFQAYVPNAN-UHFFFAOYSA-N
33589534
O=C=NC1(CCOCC1)C1=CC=CC=C1
C12H13NO2
MFCD11506354
4-isocyanato-4-phenyltetrahydropyran, 4-isocyanato-4-phenyltetrahydro-2h-pyran, 4-phenyltetrahydro-2h-pyran-4-yl isocyanate, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate
4-isocyanato-4-phenyloxane

Specifications

941717-02-8
100.0
C12H13NO2
4-isocyanato-4-phenyltetrahydropyran, 4-isocyanato-4-phenyltetrahydro-2h-pyran, 4-phenyltetrahydro-2h-pyran-4-yl isocyanate, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate
O=C=NC1(CCOCC1)C1=CC=CC=C1
203.24
203.24
4-Isocyanato-4-phenyltetrahydropyran
96.5
97%
MFCD11506354
HKKGHFQAYVPNAN-UHFFFAOYSA-N
4-isocyanato-4-phenyloxane
33589534
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is co

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.