missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxy-DL-mandelic acid, 98%, Thermo Scientific Chemicals

£29.20 - £447.00

Chemical Identifiers

CAS 55-10-7
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00004235
InChI Key CGQCWMIAEPEHNQ-UHFFFAOYSA-N
Synonym vanillylmandelic acid, vanilmandelic acid, dl-4-hydroxy-3-methoxymandelic acid, 2-hydroxy-2-4-hydroxy-3-methoxyphenyl acetic acid, 4-hydroxy-3-methoxymandelic acid, vanillomandelic acid, +/--vanillylmandelic acid, vma, alpha,4-dihydroxy-3-methoxybenzeneacetic acid, benzeneacetic acid, .alpha.,4-dihydroxy-3-methoxy
PubChem CID 1245
ChEBI CHEBI:20106
IUPAC Name 2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)C(C(=O)O)O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11490973
View Documents
Thermo Scientific Alfa Aesar
A13349.03
1 g
£29.20
1g
Please to purchase this item. Need a web account? Register with us today!
11400983
View Documents
Thermo Scientific Alfa Aesar
A13349.06
5 g
£104.00
5g
Please to purchase this item. Need a web account? Register with us today!
11410983
View Documents
Thermo Scientific Alfa Aesar
A13349.14
25 g
£447.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

55-10-7
198.174
CGQCWMIAEPEHNQ-UHFFFAOYSA-N
1245
2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
C9H10O5
MFCD00004235
vanillylmandelic acid, vanilmandelic acid, dl-4-hydroxy-3-methoxymandelic acid, 2-hydroxy-2-4-hydroxy-3-methoxyphenyl acetic acid, 4-hydroxy-3-methoxymandelic acid, vanillomandelic acid, +/--vanillylmandelic acid, vma, alpha,4-dihydroxy-3-methoxybenzeneacetic acid, benzeneacetic acid, .alpha.,4-dihydroxy-3-methoxy
CHEBI:20106
COC1=C(C=CC(=C1)C(C(=O)O)O)O

Specifications

55-10-7
C9H10O5
vanillylmandelic acid, vanilmandelic acid, dl-4-hydroxy-3-methoxymandelic acid, 2-hydroxy-2-4-hydroxy-3-methoxyphenyl acetic acid, 4-hydroxy-3-methoxymandelic acid, vanillomandelic acid, +/--vanillylmandelic acid, vma, alpha,4-dihydroxy-3-methoxybenzeneacetic acid, benzeneacetic acid, .alpha.,4-dihydroxy-3-methoxy
COC1=C(C=CC(=C1)C(C(=O)O)O)O
198.174
CHEBI:20106
98%
133°C to 134°C (decomposition)
MFCD00004235
CGQCWMIAEPEHNQ-UHFFFAOYSA-N
2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
1245
198.17
4-Hydroxy-3-methoxy-DL-mandelic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 200-224-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.