Learn More
4'-Fluoro-3'-nitroacetophenone, 98%, Thermo Scientific™
£21.51 - £50.35
Chemical Identifiers
CAS | 400-93-1 |
---|---|
Molecular Formula | C8H6FNO3 |
Molecular Weight (g/mol) | 183.138 |
MDL Number | MFCD00115369 |
InChI Key | PTCNZDJJIOLIKQ-UHFFFAOYSA-N |
Synonym | 4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone |
PubChem CID | 2734612 |
IUPAC Name | 1-(4-fluoro-3-nitrophenyl)ethanone |
SMILES | CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-] |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
400-93-1 | |
183.138 | |
PTCNZDJJIOLIKQ-UHFFFAOYSA-N | |
2734612 | |
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-] |
C8H6FNO3 | |
MFCD00115369 | |
4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone | |
1-(4-fluoro-3-nitrophenyl)ethanone |
Especificaciones
400-93-1 | |
118°C to 120°C (0.8 mmHg) | |
Odorless | |
MFCD00115369 | |
4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone | |
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-] | |
183.138 | |
183.14 | |
4'-Fluoro-3'-nitroacetophenone |
45°C to 50°C | |
>110°C (230°F) | |
C8H6FNO3 | |
2211976 | |
PTCNZDJJIOLIKQ-UHFFFAOYSA-N | |
1-(4-fluoro-3-nitrophenyl)ethanone | |
2734612 | |
98% |
Seguridad y manipulación
GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.