missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylcyclohexanone, 99%, Thermo Scientific Chemicals

Product Code. 11337376
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5441-51-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00001644
InChI Key OKSDJGWHKXFVME-UHFFFAOYSA-N
Synonym 4-ethylcyclohexanone, cyclohexanone, 4-ethyl, 4-ethyl cyclohexanone, 4-ethyl-cyclohexanone, 1-ethyl-4-oxocyclohexane, trans-4-ethylcyclohexanone, acmc-1aru9, ksc273s8l, 5-ethyl-2-oxocyclohexyl
PubChem CID 79506
IUPAC Name 4-ethylcyclohexan-1-one
SMILES CCC1CCC(=O)CC1

Specifications

Density 0.895
Boiling Point 192°C to 194°C
Flash Point 63°C (145°F)
Odor Characteristic
Refractive Index 1.4515
Quantity 25 g
Beilstein 1924964
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 4-Ethylcyclohexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.