missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-methylbenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15400058
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-08-0
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD06659870
InChI Key GAFSYSHEDOIUSI-UHFFFAOYSA-N
Synonym 4-ethoxy-3-methyl benzeneboronic acid, 4-ethoxy-3-methylbenzeneboronic acid, 4-ethoxy-3-methylphenyl boronic acid, 4-ethoxy-3-methylphenylboronicacid, boronic acid,b-4-ethoxy-3-methylphenyl, akos brn-0800, 4-ethoxy-3-methyl-phenyl boronic acid
PubChem CID 3343227
IUPAC Name (4-ethoxy-3-methylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OCC)C)(O)O

Specifications

Melting Point 168°C to 172°C
Quantity 250 mg
Formula Weight 180.01
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-3-methylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'